
1,2-Bis(2-iodoethoxy)ethane 97% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:36839-55-1
:
Formula:C₆H₁₂I₂O₂
MDL Number:MFCD00075008
:369.97 g/mol
Density:2.028 g/cm³
Flash Pt:113 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear yellow to orange-red Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
- Stabilizer:copper