Taurodeoxycholic acid sodium salt hydrate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:207737-97-1
:
Formula:C₂₆H₄₄NNaO₆S.xH₂O
MDL Number:MFCD00149238
:521.69 g/mol (anhy)
Melting Pt:204…208 °C
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Water:≤7.0 % (K.F.)
- Solubility:(5 % in water) Clear colorless to light yellow
- HPLC:≥96.0 %
- Specific optical rotation:+34.5° to +36.5° (20°C, 589 nm) (c=2.5, water) (On dry substance)
- Chloride (Cl):≤0.5 %
- Heavy metals (as Pb):≤20 ppm