Potassium citrate monohydrate 99%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6100-05-6
:
Formula:HOC(COOK)(CH₂COOK)₂·H₂O
MDL Number:MFCD00150442
Melting Pt:180 °C
Density:1,98 g/cm³ (20 °C)
:324.41 g/mol
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to white Powder or crystals
- Infrared spectrum:Conforms
- Water:4 to 6 %
- Acidity/alkalinity:Passes test
- Titration with HClO4:≥98.5 %
- Heavy metals (as Pb):≤0.001 %