In-line filters, Upchurch Scientific®
Leverantör: UPCHURCH SCIENTIFIC
Certifikat
Om denna produkt
Placed between pump and sample injection valve to trap particles released through normal piston seal wear.
- For 1/16" OD tubing
- Pressure rated to 6000 psi (414 bar)