Order Entry
Sweden
ContactUsLinkComponent
Tetramethylsilane 99%, AcroSeal®
Flammable
Tetramethylsilane 99%, AcroSeal®
Catalog # 427331000.
CAS Number:  
undefined
Flammable
Tetramethylsilane 99%, AcroSeal®
Catalog # 427331000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3575 to 1.3595 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

About this item

 :
 :88.22 g/mol
Boiling Pt:26.6 °C (1013 hPa)
Melting Pt:–99 °C
Density:0.64 g/cm³
Flash Pt:–27 °C
Storage Temperature:Refrigerator
UN:2749
ADR:3,I
 :GHS02
CAS Number:75-76-3
Formula:C₄H₁₂Si
MDL Number:MFCD00008274