Thiomorpholine 97%
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:123-90-0
- Formula:C₄H₉NS
- MDL Number:MFCD00005974
- Molecular Weight:103.19 g/mol
- Density:1.088 g/cm³
- Flash Pt:63 °C
- Storage Temperature:Ambient
- UN:3267
- ADR:8,III
2 Options Available Below
Product Details & Documents
- CAS Number:123-90-0
- Formula:C₄H₉NS
- MDL Number:MFCD00005974
- Molecular Weight:103.19 g/mol
- Density:1.088 g/cm³
- Flash Pt:63 °C
- Storage Temperature:Ambient
- UN:3267
- ADR:8,III
Documents
ACRO18797
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5370 to 1.5390 (20°C, 589 nm)
- GC:≥96.0 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



