
Hydroxyacetone, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:116-09-6
:
Formula:C₃H₆O₂
MDL Number:MFCD00004669
:74.08 g/mol
Boiling Pt:145 °C (1013 hPa)
Melting Pt:–17 °C
Density:1.08 g/cm³
Flash Pt:56 °C
Storage Temperature:Refrigerator
UN:1224
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- GC:≥88 %