Order Entry
Sweden
ContactUsLinkComponent
Solvent Blue 38, pure
Solvent Blue 38, pure
Certificates
undefined
Solvent Blue 38, pure

Specification Test Results

  • Appearance:
    Blue Powder
  • Infrared spectrum:
    Conforms
  • UV spectrum:
    Complies

Specification Test Results

Related Information