

Cyclohexylamine hydrochloride 99%
Catalog # ACRO134655000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Förpackningstyp:Plastic bottle
- Förp.:500 g
- Seq:3645
Specifications
Specification Test Results
- Appearance:White to beige Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:≥207 °C
- Water:≤0.5 % (K.F.)
- Titration with HClO4:98.5 to 101.5 %
Specification Test Results
About this item
:
Boiling Pt:306 °C (760 mmHg)
Melting Pt:207 °C
:135.64 g/mol
:GHS07
:GHS08
Storage Temperature:Omgivande
CAS Number:4998-76-9
Formula:C₆H₁₃N
MDL Number:MFCD00050540