Order Entry
Sweden
ContactUsLinkComponent
 
 
Serotonin creatinine sulphate monohydrate 99%
 
undefined
 
Serotonin creatinine sulphate monohydrate 99%

 

  • Appearance:
    Off-white to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    3.4 to 5.4 % (K.F.)
  • Sulfated ash:
    ≤0.05 %
  • Titration with NaOH:
    ≥98.5 %