(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7524-50-7
:
Formula:C₁₀H₁₄ClNO₂
MDL Number:MFCD00012489
:215.68 g/mol
Melting Pt:156…160 °C
Storage Temperature:Omgivande
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:156 to 160 °C
- Titration Argentometric:≥97.5 %
- Specific optical rotation:+37° to +39° (20 °C, 589 nm) (c=2, C2H5OH)