Rose Bengal 85%, pure, certified
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:632-69-9
:MFCD00005043
Formula:C₂₀H₂Cl₄I₄O₅Na₂
:1017.63 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Red-brown Powder
- Infrared spectrum:Conforms
- Dye content:≥80 %
- UV Lambda max:544 to 550 nm
- Absorption Ratio P-15/P+15:1.21 to 1.95
- BSC Certification:Passes