Lauroyl peroxide 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:105-74-8
:
Formula:(C₁₁H₂₃CO)₂O₂
MDL Number:MFCD00008964
:398.628 g/mol
Melting Pt:53…57 °C
Storage Temperature:Refrigerator
UN:3106
ADR:5.2,
:GHS02
Specification Test Results
- Appearance:White Powder or granules or flakes or pellets
- Infrared spectrum:Conforms
- Melting point:53 to 57 °C
- Titration Iodometric:≥98.5 %