
Cresyl violet (acetate), pure, high purity biological stain
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10510-54-0
:
Formula:C₁₈H₁₅N₃O₃
MDL Number:MFCD00013151
:321.33 g/mol
Melting Pt:140…143 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark green to black Crystalline powder
- Infrared spectrum:Conforms
- Dye content:≥65 %