Sterilin™ Tubings, Silicone
Leverantör: STERILIN
Certifikat
Om denna produkt
Sterilin™ silicone tubing is non-toxic and ideal for medical, pharmaceutical and food applications with high quality, nontoxic Thermo Scientific™ Sterilin™ Silicone Tubing. Not available in all geographic regions.
- Laser etched batch code for ease of traceability
- Ideal for use with peristaltic pumps
- Can be autoclaved or dry heat sterilised
- 15 m length
- Aseptically manufactured