Order Entry
Sweden
ContactUsLinkComponent
Desiccator lids
Desiccator lids
Katalog # NALG71-5311-0250
   Low Manufacturing Impact
undefined
Desiccator lids
Katalog # NALG71-5311-0250
   Low Manufacturing Impact
Specifikationer
  • Type:
    With recessed rim
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Pk:
    2
Specifikationer
Om denna produkt
Transparent, lightweight, and unbreakable polycarbonate