Anti-WNK1 Antibody
Catalog # BSBTPB9964
Supplier: Boster Bio
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:WNK lysine deficient protein kinase 1
- Antigensymbol:WNK1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- ImmunKemi:Yes
- Isotyp:IgG
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Korsad adsorption:No
- Gen-ID:65125
- Antigen synonymer:PSK|KDP|HSAN2|p65|PPP1R167|HSN2|PRKWNK1
- Storlek:100 µg
- Förp.:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Serine/threonine-protein kinase WNK1(WNK1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: WNK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat