Anti-ISG15 Antibody
Catalog # BSBTPB9951
Supplier: Boster Bio
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:ISG15 ubiquitin-like modifier
- Antigensymbol:ISG15
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- ImmunKemi:Yes
- Isotyp:IgG
- Reaktivitet:Mouse
- Western blot:Yes
- Korsad adsorption:No
- Gen-ID:100038882
- Antigen synonymer:IGI15|IP17|Irfp|UCRP|G1p2
- Storlek:100 µg
- Förp.:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Ubiquitin-like protein ISG15(ISG15) detection. Tested with WB, IHC-P in Mouse.
Type: Primary
Antigen: ISG15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse