Order Entry
Sweden
ContactUsLinkComponent
 
Anti-ISG15 Antibody
Catalog # BSBTPB9950
Supplier:  Boster Bio
undefined
Anti-ISG15 Antibody
Catalog # BSBTPB9950
Supplier:  Boster Bio

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    ISG15 ubiquitin-like modifier
  • Antigensymbol:
    ISG15
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Gen-ID:
    9636
  • Antigen synonymer:
    IP17|hUCRP|IMD38|UCRP|IFI15|G1P2
  • Storlek:
    100 µg
  • Förp.:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Ubiquitin-like protein ISG15(ISG15) detection. Tested with WB, IHC-P in Human.

Type: Primary
Antigen: ISG15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human