Anti-TCP1 epsilon Antibody
BSBTPB9928
- Antibody type:Primary
- Antigen name:chaperonin containing TCP1, subunit 5 (epsilon)
- Antigen symbol:CCT5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:22948
- Antigen synonyms:CCTE|TCP-1-epsilon|CCT-epsilon|PNAS-102|HEL-S-69
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for T-complex protein 1 subunit epsilon(CCT5) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: CCT5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat