Anti-Osteocalcin Antibody
Catalog # BSBTPB9920
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:bone gamma-carboxyglutamate (gla) protein
- Antigen symbol:BGLAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat
- Cross adsorption:No
- Gene ID:25295
- Antigen synonyms:Bgp|Bgpr|Bglap2|Bgpra
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Osteocalcin(BGLAP) detection. Tested with IHC-P in Rat.
Type: Primary
Antigen: BGLAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat