Order Entry
Sweden
ContactUsLinkComponent
Anti-Osteocalcin Antibody
Anti-Osteocalcin Antibody
  BSBTPB9918
undefined
Anti-Osteocalcin Antibody
  BSBTPB9918

 

  • Antibody type:
    Primary
  • Antigen name:
    bone gamma-carboxyglutamate (gla) protein
  • Antigen symbol:
    BGLAP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Gene ID:
    632
  • Antigen synonyms:
    OCN|OC|BGP
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Osteocalcin(BGLAP) detection. Tested with IHC-P in Human.

Type: Primary
Antigen: BGLAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human