Contrast clips
Supplier: witeg Labortechnik
Certificates
About this item
PP, white/black.
- For attachment to all measuring flasks
- With grip deepening
- Unbreakable, resistant to acids and alkalis
- For better recognition and adjustment of the liquid level