Calcium stearate NF
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:1592-23-0
:66071-81-6
Formula:C₃₆H₇₀CaO₄
MDL Number:MFCD00036390
:607.03 g/mol
Merck Index:13,01708
Melting Pt:147…149 °C
Density:1.065 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:216-472-8
Specification Test Results
- Assay:9% Min.