Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cysteaminium chloride
Cysteaminium chloride
Cysteaminium chloride
Cysteaminium chloride
Supplier:
Spectrum Chemical
Certificates
Expand image
Cysteaminium chloride
Supplier:
Spectrum Chemical
Certificates
About this item
CAS Number:
156-57-0
:
Formula:
C₂H₇NS·HCl
MDL Number:
MFCD00012904
:
113.61 g/mol
EINECS:
205-858-1
Specification Test Results
Assay:
98% Min.
Specification Test Results
Related Information
Documentation
Certificates
Search by Lot Number
Documentation