Brij® L23
Catalog # SPCMBR105-12KG
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Poly pail
- Pk:12 kg
- Seq:4187
Specifications
About this item
:
EINECS:500-002-6
:GHS07
Storage Temperature:Ambient
CAS Number:9002-92-0
Formula:(C₂₀H₄₂O₅)n
MDL Number:MFCD00043063