Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Agar
Agar, powder NF
Agar, powder NF
Agar, powder NF
Supplier:
Spectrum Chemical
Certificates
Expand image
Agar, powder NF
Supplier:
Spectrum Chemical
About this item
CAS Number:
9002-18-0
:
MDL Number:
MFCD00081288
Merck Index:
13,00184
Melting Pt:
90 °C
Storage Temperature:
Ambient
EINECS:
232-658-1