Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L(-)-Menthol
L(-)-Menthol, crystals USP
L(-)-Menthol, crystals USP
:
Spectrum Chemical
L(-)-Menthol, crystals USP
:
Spectrum Chemical
:
2216-51-5
:
:
C₁₀H₂₀O
:
MFCD00062979
:
156.27 g/mol
:
13,05861
:
212 °C (1013 hPa)
:
41…44 °C
:
0.89 g/cm³ (20 °C)
:
96 °C
:
218-690-9
:
GHS07
:
Ambient