3-Aminophenol
Poly pail, 10 kg
Catalog # SPCMA2349-10KGBL
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Synonyms: 3-Hydroxyaniline, m-Aminophenol
- CAS Number:591-27-5
- Formula:C₆H₇NO
- MDL Number:MFCD00007786
- Molecular Weight:109.13 g/mol
- Merck Index:13,00459
- Boiling Pt:299 °C (1013 hPa)
- Melting Pt:122 °C
- Density:1.275 g/cm³ (20 °C)
- Flash Pt:92 °C
- EINECS:209-711-2
- UN:2512
- ADR:6.1,III
Specifications
Specifications
- Pack type:Poly pail
- Pk:10 kg
- Cat. No.:SPCMA2349-10KGBL



