Order Entry
Sweden
Orders LinkContactUsLinkComponent
Diethylene glycol

Diethylene glycol

Irritant
Supplier:   Spectrum Chemical
Diethylene glycol
Supplier:  Spectrum Chemical
Certificates
Diethylene glycol

2 Options Available Below

Product Details & Documents
Synonyms:  2,2'-Oxybis(ethan-1-ol), Digol, 2,2'-Oxybisethanol
  • CAS Number:111-46-6
  • Formula:C₄H₁₀O₃
  • MDL Number:MFCD00002882
  • Molecular Weight:106.12 g/mol
  • Merck Index:13,03146
  • Boiling Pt:245 °C (1013 hPa)
  • Melting Pt:–10 °C
  • Density:1.118 g/cm³ (20 °C)
  • Flash Pt:123 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:203-872-2
Specifications for Product Family

Specification Test Results

  • Assay:
    97% Min.
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Poly drum
    20 L
    Log in for availability
    Log in for availability
    1 * 20 L
    1 * 20 L

  • 200 L
    Log in for availability
    Log in for availability
    1 * 200 L
    1 * 200 L - 26 375,00 kr
    26 375,00 kr
    1 * 200 L
       26 375,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.