Adenine (synthetic)
Catalog # SPCMAD105-1KGBL
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Glass, amber
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:99% Min.
Specification Test Results
About this item
:
:135.13 g/mol
Merck Index:13,00152
Density:0.99741 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:200-796-1
UN:2811
ADR:6.1,III
:GHS06
CAS Number:73-24-5
Formula:C₅H₅N₅
MDL Number:MFCD00041790