
2-(4-Chlorophenyl)propionic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:938-95-4
:
Formula:C₉H₉ClO₂
MDL Number:MFCD00044670
:184.62 g/mol
Boiling Pt:121…126 °C (0.2 mmHg)
Melting Pt:55…59 °C
Storage Temperature:Ambient
EINECS:213-351-1
:GHS07