Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Fusidate sodium

Fusidate sodium
Certificates
Fusidate sodium

2 Options Available Below

Product Details & Documents

CAS No.: 751-94-0 Molecular Formula: C31H47NaO6 Formula Weight: 538.69 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Moisture and Light Sensitive Appearance: White to off-white Solubility: Soluble in water MDL No.: MFCD09054714

Applications: Targets gram positive bacteria namely those which cause skin infections

Synonyms:  Fusidate Sodium, Fusidic acid sodium, Sodium (Z)-2-((3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetoxy-3,11-dihydroxy-4,8,10,14-tetramethyldodecahydro-1H-cyclopenta[a]phenanthren-17(2H,10H,14H)-ylidene)-6-methylhept-5-enoate
  • CAS Number:751-94-0
  • Formula:C₃₁H₄₇NaO₆
  • MDL Number:MFCD09054714
  • Molecular Weight:538.701 g/mol
  • Merck Index:14,4317
  • EINECS:212-030-3
Product Family Options

Product Information