Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
MIF antagonist, ISO-1
MIF antagonist, ISO-1
MIF antagonist, ISO-1
MIF antagonist, ISO-1
Catalog # J64947.MB
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
Certificates
Expand image
MIF antagonist, ISO-1
Catalog # J64947.MB
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
Certificates
Specifications
Pk:
25 mg
Seq:
2315
Specifications
About this item
Product Details
Documentation
:
:
235.2 g/mol
CAS Number:
478336-92-4
Formula:
C₁₂H₁₃NO₄
MDL Number:
MFCD22376813
Product Details
Documentation