Trimyristin 95%
Catalog # J62819.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 g
Specifications
About this item
CAS Number:555-45-3
:
Formula:C₄₅H₈₆O₆
MDL Number:MFCD00036229
Merck Index:14,9724
Melting Pt:56…57 °C
:723.18 g/mol
EINECS:209-099-7