Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Paraformaldehyde
Paraformaldehyde fixative solution
Paraformaldehyde fixative solution
Health
Irritant
Paraformaldehyde fixative solution
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Health
Irritant
Paraformaldehyde fixative solution
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
30525-89-4
:
Formula:
(CH₂O)n
MDL Number:
MFCD00133991
:
GHS08
:
GHS07
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAJ61984
Videos
Thermo Scientific Acroseal 25 Years
Documentation