Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(1-Hydroxyethyl)pyridine
4-(1-Hydroxyethyl)pyridine 97%
4-(1-Hydroxyethyl)pyridine 97%
4-(1-Hydroxyethyl)pyridine 97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
4-(1-Hydroxyethyl)pyridine 97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
23389-75-5
:
Formula:
C₇H₉NO
MDL Number:
MFCD00077867
Melting Pt:
51…55 °C
:
123.15 g/mol
:
GHS07
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH58711
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation