Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
S-Phenyl-L-cysteine
S-Phenyl-L-cysteine 97%
S-Phenyl-L-cysteine 97%
S-Phenyl-L-cysteine 97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
S-Phenyl-L-cysteine 97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
34317-61-8
:
Formula:
C₉H₁₁NO₂S
MDL Number:
MFCD01318758
:
197.252 g/mol
Melting Pt:
200 °C (dec.)
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH55031
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation