0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
4-Ethynylpyridine hydrochloride
4-Ethynylpyridine hydrochloride 97%
4-Ethynylpyridine hydrochloride 97%
:
Thermo Fisher Scientific
4-Ethynylpyridine hydrochloride 97%
:
Thermo Fisher Scientific
:
352530-29-1
:
:
C₇H₆ClN
:
MFCD03093912
:
139.58 g/mol
:
∼ 150 °C (dec.)
:
GHS07
:
Ambient
msds_ALFAH53455
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years