Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(4-Aminopyridin-3-yl)methanol
(4-Aminopyridin-3-yl)methanol 95%
(4-Aminopyridin-3-yl)methanol 95%
Toxic
Irritant
(4-Aminopyridin-3-yl)methanol 95%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Toxic
Irritant
(4-Aminopyridin-3-yl)methanol 95%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
138116-34-4
:
Formula:
C₆H₈N₂O
MDL Number:
MFCD06203066
:
124.14 g/mol
Melting Pt:
167…169 °C
UN:
2811
ADR:
6.1,III
:
GHS06
:
GHS07
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH50033
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation