



3-Methyl-2-butenal 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:107-86-8
:
Formula:C₅H₈O
MDL Number:MFCD00010291
:84.12 g/mol
Merck Index:14,8448
Boiling Pt:136 °C
Melting Pt:–20 °C
Density:0,88
Flash Pt:37 °C (99 °F)
EINECS:203-527-6
ADR:6.1,II
:GHS06
:GHS05
:GHS02
:GHS07