Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Allyl-1,6-heptadien-4-ol
4-Allyl-1,6-heptadien-4-ol 99%
4-Allyl-1,6-heptadien-4-ol 99%
Irritant
4-Allyl-1,6-heptadien-4-ol 99%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
4-Allyl-1,6-heptadien-4-ol 99%
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
10202-75-2
:
Formula:
(CH₂=CHCH₂)₃COH
MDL Number:
MFCD00039829
:
152.24 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
SDS
msds_ALFAB20703
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation