0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Ethyl-2-oxo-4-phenylbutyrate
Ethyl-2-oxo-4-phenylbutyrate ≥90%
Ethyl-2-oxo-4-phenylbutyrate ≥90%
:
Thermo Fisher Scientific
Ethyl-2-oxo-4-phenylbutyrate ≥90%
:
Thermo Fisher Scientific
:
64920-29-2
:
:
C₁₂H₁₄O₃
:
MFCD00037533
:
131…133 °C (2 mmHg)
:
1.09
:
>110 °C (230 °F)
:
206.24 g/mol
:
Ambient
:
265-276-9
msds_ALFAB20148
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years