Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Palladium
Palladium 1% on activated carbon powder wetted with ca. 50% water, reduced
Palladium 1% on activated carbon powder wetted with ca. 50% water, reduced
Palladium 1% on activated carbon powder wetted with ca. 50% water, reduced
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Palladium 1% on activated carbon powder wetted with ca. 50% water, reduced
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
7440-05-3
:
Formula:
Pd
MDL Number:
MFCD00011167
:
106.420 g/mol
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA038292
Videos
Thermo Scientific Acroseal 25 Years
Documentation