Anti-BOD1L1 Goat Polyclonal Antibody
Catalog # EVBIEB11986
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:biorientation of chromosomes in cell division 1-like 1
- Antigen symbol:BOD1L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Format:affinity purified
- Form:liquid
- Gene ID:259282
- Antigen synonyms:BOD1L|FAM44A
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Sequence:CPETEPHDTKEENS
- Storage temperature:Store at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing
- Concentration:100µg/200µl
- Shipping temperature:8 °C
- Purification:Affinity purified
- Size:100µg
- Pack type:plastic vial
- Pk:0,1 mg
Specifications
About this item
Western Blot: Not yet tested - our routinely used western blotting protocol does not allow detection of proteins as large as the calculated size of 330kDa according to NP_683692.2. Therefore we cannot recommend an optimal concentration and the antibody i
Type: Primary
Antigen: BOD1L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse