Anti-POLB Rabbit Polyclonal Antibody
Catalog # BSBTPB9812
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Polymerase (DNA directed), beta
- Antigen symbol:POLB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5423
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: POLB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human