Anti-EME1 Rabbit Polyclonal Antibody
BSBTPB9804
- Antibody type:Primary
- Antigen name:Essential meiotic structure-specific endonuclease 1
- Antigen symbol:EME1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Gene ID:146956
- Antigen synonyms:MMS4L|SLX2A
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: EME1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat