Order Entry
Sweden
ContactUsLinkComponent
Anti-HDAC1 Rabbit Polyclonal Antibody
Anti-HDAC1 Rabbit Polyclonal Antibody
Catalog # ABGEAP16867C
Supplier:  Abgent
Anti-HDAC1 Rabbit Polyclonal Antibody
Catalog # ABGEAP16867C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Histone deacetylase 1
  • Antigen symbol:
    HDAC1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    internal
  • Gene ID:
    3065
  • Antigen synonyms:
    GON-10|RPD3L1|RPD3|HD1
  • Amino acid number:
    402 to 430
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    55.103 kDa
  • Sequence:
    CEEE(FS)DSEEEGEGG
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.45
  • Shipping temperature:
    4 °C
  • Immunogen:
    This HDAC1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 402-430 amino acids from the Central region of human HDAC1.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: HDAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat