Order Entry
Sweden
ContactUsLinkComponent
Anti-CTGF Mouse Monoclonal Antibody [clone: 558CT34.4.4]
Anti-CTGF Mouse Monoclonal Antibody [clone: 558CT34.4.4]
  ABGEAM2095B
 :  Abgent
Anti-CTGF Mouse Monoclonal Antibody [clone: 558CT34.4.4]
  ABGEAM2095B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    connective tissue growth factor
  • Antigen symbol:
    CTGF
  • Clonality:
    Monoclonal
  • Clone:
    558CT34.4.4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    1490
  • Antigen synonyms:
    IGFBP8|HCS24|CCN2|NOV2
  • Amino acid number:
    166 to 193
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    38.091 kDa
  • Sequence:
    AYRLEDTFGPDPTC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    1.200
  • Shipping temperature:
    4 °C
  • Immunogen:
    This CTGF antibody is generated from mice immunized with a KLH conjugated synthetic peptide between 166-193 amino acids from human CTGF.
  • Purification:
    Protein G Purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: CTGF
Clonality: Monoclonal
Clone: 558CT34.4.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human