Order Entry
Sweden
ContactUsLinkComponent
Anti-NFKB1 Mouse Monoclonal Antibody [clone: 1298CT792.105.117.133]
Anti-NFKB1 Mouse Monoclonal Antibody [clone: 1298CT792.105.117.133]
Catalog # ABGEAM2254A
Leverantör:  Abgent
Anti-NFKB1 Mouse Monoclonal Antibody [clone: 1298CT792.105.117.133]
Catalog # ABGEAM2254A
Leverantör:  Abgent

Specifikationer

  • Antibody type:
    Primary
  • Antigen name:
    Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1
  • Antigen symbol:
    NFKB1
  • Clonality:
    Monoclonal
  • Clone:
    1298CT792.105.117.133
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    4790
  • Antigen synonyms:
    NFkappaB|EBP-1|p50|NFKB-p50|NFKB-p105|NF-kB1|NF-kappaB|p105|KBF1|NF-kappa-B
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    105.356 kDa
  • Sequence:
    MAEDDPYLGR PEQMFHLDPS LTHTIFNPEV FQPQMALPTD GPYLQILEQP KQRGFRFRYVCEGPSHGGLP GASSEKNKKS YPQVKICNYV GPAKVIVQLV TNGKNIHLHA HSLVGKHCEDGICTVTAGPK DMVVGFANLG ILHVTKKKVF ETLEARMTEA CIRGYNPGLL VHPDLAYLQAEGGGDRQLGD REKELIRQAA LQQTKEMDLS
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.5
  • Shipping temperature:
    4 °C
  • Immunogen:
    This antibody is generated from a mouse immunized with a recombinant protein from human NFKB1.
  • Purification:
    Protein G Purification
  • Pk:
    400 µl

Specifikationer

Om denna produkt

Type: Primary
Antigen: NFKB1
Clonality: Monoclonal
Clone: 1298CT792.105.117.133
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human